missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2B6 Antibody (OTI3D5), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70526R
This item is not returnable.
View return policy
Description
Cytochrome P450 2B6 Monoclonal specifically detects Cytochrome P450 2B6 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Cytochrome P450 2B6 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CYP2B6 | |
| Full length recombinant protein of human Cytochrome P450 2B6 (NP_000758) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3D5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CPB6, CYP2B, CYP2B7, CYP2B7P, CYPIIB6cytochrome P450, subfamily IIB (phenobarbital-inducible), cytochrome P450 2B6, Cytochrome P450 IIB1, cytochrome P450, family 2, subfamily B, cytochrome P450, family 2, subfamily B, polypeptide 6, cytochrome P450, subfamily IIB (phenobarbital-inducible), polypeptide 6, EC 1.14.14.1, IIB1, P450 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 1555.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction