missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP14C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-33483
This item is not returnable.
View return policy
Description
UTP14C Polyclonal specifically detects UTP14C in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| UTP14C | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Q5TAP6 | |
| UTP14C | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ARMMERMSLKHQNSGKWAKSKAIMAKYDLEARQAMQEQLAKNKELTQKLQVASESEEE | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 2700066J21Rik, KIAA0266, U3 Small Nucleolar RNA-Associated Protein 14 Homolog C, UTP14, U3 Small Nucleolar Ribonucleoprotein, Homolog C (Yeast), UTP14B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9724 | |
| Human | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur