missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP18 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97492AF405
This item is not returnable.
View return policy
Description
UTP18 Polyclonal specifically detects UTP18 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| UTP18 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 405 | |
| CGI-48, U3 small nucleolar RNA-associated protein 18 homolog, UTP18, small subunit (SSU) processome component, homolog (yeast), WD repeat domain 50, WD repeat-containing protein 50, WDR50 | |
| Produced in rabbits immunized with E. coli-derived Human UTP18 fragment. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51096 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction