missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UTS2 Recombinant Rabbit Monoclonal Antibody (3B2M6)

Product Code. 30131587
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30131587 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30131587 Supplier Invitrogen™ Supplier No. MA555591

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen Sequence: SLPLLDSREI SFQLSAPHED ARLTPEELER ASLLQILPEM LGAERGDILR KADSSTNIFN PRGNLRKFQD FSGQDPNILL SHLLARIWKP YKKRETPDCF WKYCV.

Urotensin II is an active cyclic heptapeptide absolutely conserved from lamprey to human. The active peptide acts as a vasoconstrictor and is expressed only in brain tissue. Despite the gene family name similarity, this gene is not homologous to urocortin, a member of the sauvagine/corticotropin releasing factor/urotensin I family. Most of the proprotein is cleaved to make the mature peptide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTS2
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3B2M6
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene UTS2
Gene Accession No. Q9QZQ4
Gene Alias prepro U-II; prepro-UII; PRO1068; Ucn2; UII; U-II; UNQ525/PRO1068; urocortin 2; urotensin 2; urotensin II; Urotensin-2; urotensin-II; UTS2
Gene Symbols UTS2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 20-124 of human Urotensin II (UTS2).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29180
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.