Learn More
Vilber Lourmat™ UV Tube
Product Code. 11860250
Change view
Click to view available options
Wavelength:
254 nm
312 nm
365 nm
Wattage:
6 W
8 W
15 W
30 W
40 W
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength Wattage unitSize
11860250 312 nm 8 W Each
11796678 254 nm 30 W Each
11786688 254 nm 8 W Each
11756778 254 nm 40 W Each
11706688 365 nm 8 W Each
11716578 254 nm 6 W Each
11736588 365 nm 15 W Each
15364932 312 nm 15 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11860250 Supplier Vilber Lourmat™ Supplier No. 907520821
Only null left
Add to basket
Add to basket
Rich, robust UV (ultraviolet) tube that is ideal for use in many laboratory, research, scientific, and other environments
  • Standard diameter 16 mm
Specifications
Wattage 8 W
Wavelength 312 nm
Diameter (Metric) 16 mm
Length (Metric) 288 mm
Product Type UV Tube