Learn More
Intersafe Groeneveld™ Uvex Astrospec Safety Eyewear
Product Code. 10286124
Encompass
Change view
Click to view available options
Frame Color:
Blue
Green
Yellow
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Frame Color unitSize
10286124 Blue Each
10461544 Yellow Each
10327004 Green Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10286124 Supplier Intersafe Groeneveld™ Supplier No. 110066
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
For protection against splashes and dust.
  • Lens material: Polycarbonate
  • Supravision HC-AF surface coating
Specifications
Certifications/Compliance CE Marked
Frame Color Blue
Lens Material Polycarbonate