Learn More
Intersafe Groeneveld™ Uvex Astrospec Safety Eyewear
Product Code. 10461544
Encompass
Click to view available options
Frame Color:
Blue
Green
Yellow
Unit Size:
Each
Product Code. 10461544
Brand: Intersafe Groeneveld™ 110065
This item requires a price update prior to being ordered.
Request a Price for this item here.
For protection against splashes and dust.
  • Lens material: Polycarbonate
  • Supravision HC-AF surface coating
Specifications
Certifications/Compliance CE Marked
Frame Color Yellow
Lens Material Polycarbonate