Learn More
UVP Clear UV Blocking Covers
Specifications
Specifications
| For Use With (Application) | Gel electrophoresis |
| Color | Clear |
| For Use With (Equipment) | 3uv™ transilluminators |
| Includes | Uv Blocking Cover |
| Quantity | each |
| Size | 10.8 x 13.8 in. |
| Product Type | UV Blocking Cover |