Learn More
UVP Filter Assemblies for Handheld UV Lamps
Brand: UVP 98000403
Additional Details : Weight : 0.80000kg
Specifications
Model UVGL-58 handheld UV lamp | |
Each | |
Filter assembly | |
254nm and 365nm |