Learn More
Invitrogen™ UVRAG Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597066
Description
Immunogen sequence: KGHRKRASSE NERLQYKTPP PSYNSALAQP VTTVPSMGET ERKITSLSSS LDTSLDFSKE NKKKGEDLVG SLNGGHANVH PSQEQGEALS GHRATVNGTL LPSEQAGSAS VQLPGEFHPV SEAELCCTVE QAEEIIGLEA TGFASGDQLE AFNCIPVDSA VAVECDEQVL GEFEEFSRRI YALNENVSSF RRPRRSSDK; Positive Samples: LO2, HeLa, Mouse liver, Rat liver, Rat brain; Cellular Location: Chromosome, Early endosome, Endoplasmic reticulum, Late endosome, Lysosome, Midbody, centromere.
UVRAG is predicted to contain a coiled-coil structure and is likely to be metabolically unstable based on the occurrence of a strong PEST domain. It interacts with Beclin 1, leading to activation of autophagy and thereof inhibition of tumorigenicity of human colon cancer cells. The gene for UVRAG protein is localized on human chromosome 11q13. 5 and complements the ultraviolet sensitivity of xeroderma pigmentosum group C cells. The gene has been extremely well conserved during evolution, implying an important role for this gene product in cell metabolism. Chromosomal aberrations involving this gene are associated with left-right axis malformation, and mutations in this gene have been associated with colon cancer. The protein is highly expressed in human brain, lung, kidney and liver.
Specifications
| UVRAG | |
| Polyclonal | |
| Unconjugated | |
| Uvrag | |
| 9530039D02Rik; AI648770; BB124205; beclin 1 binding protein; DHTX; disrupted in heterotaxy; hypothetical protein; p63; UV radiation resistance associated; UV radiation resistance associated gene; UV radiation resistance-associated gene protein; UV radiation resistance-associated protein; UVRAG; Uvrag1; Uvragl; VPS38 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 308846, 7405, 78610 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8K245, Q9P2Y5 | |
| Uvrag | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 501-699 of human UVRAG (NP_0033602). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.