Learn More
Vβ4 T-Cell Receptor Rat anti-Mouse, Brilliant Violet 421, Clone: KT4, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 743019
Description
The KT4 antibody specifically recognizes the Vβ4 T-cell Receptor (TCR) of mice having the a ( e.g /., C57BR, C57L, SJL, SWR), b ( e.g /., A, AKR, BALB/c, CBA, C3H/He, C57BL, C58, DBA/1, DBA/2), and c ( e.g /., RIII) haplotypes of the Tcrb /gene complex. Vβ4 TCR-bearing cells are among the CD4+ autoreactive T cells which induce autoimmune thyroiditis after elimination of regulatory (CD5-bright) T-cell subsets. Plate-bound KT4 antibody activates Vβ4 TCR-bearing T cells.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications
| Vβ4 T-Cell Receptor | |
| KT4 | |
| Brilliant Violet 421 | |
| TCR V beta 4, TCR Vβ4 | |
| KT4 B10.D2 mouse T-cell clone I3 | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.