missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vδ 4 TCR Armenian Hamster anti-Mouse, BB700, Clone: GL2, BD OptiBuild™
Armenian Hamster Monoclonal Antibody
Brand: BD Biosciences 746149
This item is not returnable.
View return policy
Description
Vδ 4 TCR Monoclonal antibody specifically detects Vδ 4 TCR in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Vδ 4 TCR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Armenian Hamster | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| GL2 | |
| Brilliant Blue 700 (BB700) | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction