Learn More
Vδ 6.3/2 TCR Hamster anti-Mouse, Brilliant Violet 711, Clone: 8F4H7B7, BD Optibuild™
Armenian Hamster Monoclonal Antibody
Brand: BD Biosciences 744476
Description
The 8F4H7B7 antibody reacts with Vδ6.3/2 T-cell Receptor (TCR)-bearing T lymphocytes. Originally defined as a member of the Vδ6 TCR subfamily, it is now proposed that the C57BL-derived Vδ6.3 is an allelic variant of Vδ6.2, found in A/J, AKR, BALB/c, C3H/He, and FVB mice. mAb 8F4H7B7 crossreacts with Vδ6.4 and possibly Vδ6.6 in DBA/2 mice, and it also detects a subset ofγδTCR-bearing cells in CBA/J and C57L mice. It does not recognize Vδ4, Vδ5, Vδ6.1, or Vδ6.5 TCR. A subpopulation of thymocytes expressing Vδ6.3 or Vδ6.4 TCR (in C57BL/6 or DBA/2 mice, respectively) and low levels of CD90.2 (Thy-1.2) shares functional and phenotypic characteristics with NK-T cells. SimilarδV 6.4 TCRexpressing lymphocytes make up significant proportions of theγδT-cell populations in the liver and spleen of DBA/2 mice. Furthermore, T lymphocytes bearing Vδ6.3/2 TCR are found in the skin and intestinal epithelium and may represent a unique T-cell subpopulation with a potential for autoimmune reactivity.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
Specifications
| Vδ 6.3/2 TCR | |
| 8F4H7B7 | |
| Brilliant Violet 711 | |
| V delta 6.3/2 TCR, TCR Vδ 6.3/2, TCR Vδ6.3/2, TCR Vd6.3/2 | |
| 1E6.11.D7, a C57BL/6 mouse Vγ1.1Vδ6.3 TCR-bearing IEL/T-cell hybridoma | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Armenian Hamster | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.