Learn More
Vβ 6 T-Cell Receptor Rat anti-Mouse, Brilliant Violet 421, Clone: RR4-7, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 744590
Description
The RR4-7 antibody specifically reacts with the V β 6 T-Cell Receptor (TCR) of mice having the a ( e.g., C57BR, C57L, SJL) and b ( e.g., A, BALB/c, CBA/Ca, C3H/He, C57BL, DBA/1) haplotypes of the Tcrb gene complex. The Tcrb-V6 gene locus is deleted in mice having the c ( e.g., RIII) haplotype. V β 6 TCR-bearing T lymphocytes are clonally eliminated in mice expressing superantigen encoded by Mtv-7 ( Mls-1[a], Mls[a] ) endogenous provirus ( e.g., AKR, CBA/J, C58, DBA/2, NZB), or Mtv -43 endogenous provirus ( e.g., MA/MyJ). Exogenous MMTV-SW, as well as endogenous Mtv -44-encoded superantigen ( e.g., NZW), also causes incomplete elimination of V β 6 TCR-expressing T cells. Plate-bound RR4-7 antibody activates V β 6 TCR-bearing T cells, soluble RR4-7 mAb blocks in vitro proliferation and cytolytic activities of V β 6 TCR-bearing T-cell clones, and injection of the antibody results in in vivo depletion of V β 6 TCR-bearing T cells.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications
| Vβ 6 T-Cell Receptor | |
| RR4-7 | |
| Brilliant Violet 421 | |
| TCR V beta 6, TCR Vβ6 | |
| C57BL/6 Mouse Helper T-Cell Clone OI11 | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2b λ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.