missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ V-ATPase E1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16300035
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16300035 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16300035 Supplier Invitrogen™ Supplier No. PA583071

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KEKQIEQQKK IQMSNLMNQA RLKVLRARDD LITDLLNEAK QRLSKVVKDT TRYQVLLDGL VLQGLYQLLE PRMIVRCRKQ DFPLVKAAVQ KAIPMYKIAT KNDVDVQIDQ ESYLPEDIAG GVEIYNGDRK IKVSNTLESR LDLIAQQMMP EVRGALFGAN ANRKF Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 99%.

Subunit of the peripheral V1 complex of vacuolar ATPase essential for assembly or catalytic function. V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen V-ATPase E1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP6V1E1
Gene Accession No. P36543
Gene Alias 2410029D23Rik; Atp6e; Atp6e2; ATP6V1E; Atp6v1e1; ATPase H+ transporting V1 subunit E1; ATPase, H+ transporting lysosomal (vacuolar proton pump), 32 kDa; ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E; ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E1; ATPase, H+ transporting, lysosomal V1 subunit E1; ATPase, H+ transporting, V1 subunit E; D6Ertd385e; E2; H(+)-ATPase E-like protein; H(+)-transporting two-sector ATPase, 31kDa subunit; H+ ATPase subunit E; H+-transporting ATP synthase chain E, vacuolar; lysosomal 31kDa; P31; vacuolar H(+)-ATPase, E subunit; vacuolar H+ ATPase E1; vacuolar proton pump subunit E 1; V-ATPase 31 kDa subunit; V-ATPase subunit E; V-ATPase subunit E 1; VATPase, H+ transporting, lysosomal V1 subunit E1; V-ATPase, subunit E; Vma4; V-type proton ATPase subunit E 1
Gene Symbols ATP6V1E1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP6V1E1. Recombinant protein control fragment (Product #RP-91321).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 529
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.