missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ V-ATPase H Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA554793

Product Code. 15752721

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PQVLAVAAHD VGEYVRHYPR GKRVIEQLGG KQLVMNHMHH EDQQVRYNAL LAVQKLMVHN WEYLGKQLQS E Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular organelles. V-ATPase-dependent organelle acidification is necessary for multiple processes including protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. The encoded protein is the regulatory H subunit of the V1 domain of V-ATPase, which is required for catalysis of ATP but not the assembly of V-ATPase. Decreased expression of this gene may play a role in the development of type 2 diabetes. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

V-ATPase H
Polyclonal
Unconjugated
ATP6V1H
0710001F19Rik; Atp6v1h; ATPase H+ transporting V1 subunit H; ATPase, H+ transporting, lysosomal 50/57kDa, V1 subunit H; ATPase, H+ transporting, lysosomal V1 subunit H; AU022349; CGI-11; MSTP042; NBP1; Nef-binding protein 1; protein VMA13 homolog; SFD; SFDalpha; SFDbeta; vacuolar ATP synthase subunit H; vacuolar ATPase subunit H; vacuolar proton pump H subunit; vacuolar proton pump subunit H; Vacuolar proton pump subunit SFD; V-ATPase 50/57 kDa subunits; V-ATPase H subunit; V-ATPase subunit H; VMA13; V-type proton ATPase subunit H
Rabbit
Antigen Affinity Chromatography
RUO
51606
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9UI12
ATP6V1H
Recombinant protein corresponding to Human V-ATPase H. Recombinant protein control fragment (Product #RP-93733).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.