missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VABP Antibody (6C239.4), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24876IR
This item is not returnable.
View return policy
Description
VABP Monoclonal specifically detects VABP in Human samples. It is validated for Western Blot.
Specifications
| VABP | |
| Monoclonal | |
| DyLight 755 | |
| Q9NZI2 | |
| KCNIP1 | |
| Full length recombinant protein was used as the immunogen. | |
| 0.1 mL | |
| Primary | |
| This antibody is reactive to Human. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot | |
| 6C239.4 | |
| Western Blot | |
| A-type potassium channel modulatory protein 1, KChIP1, KCHIP1MGC95, Kv channel interacting protein 1, Kv channel-interacting protein 1, potassium channel interacting protein 1, Potassium channel-interacting protein 1, VABP, Vesicle APC-binding protein | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 30820 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction