missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAC14 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44278AF647
This item is not returnable.
View return policy
Description
VAC14 Polyclonal antibody specifically detects VAC14 in Human samples. It is validated for Western Blot
Specifications
| VAC14 | |
| Polyclonal | |
| Western Blot | |
| ArPIKfyve, FLJ10305, FLJ36622, MGC149816, protein VAC14 homolog, Tax1 (human T-cell leukemia virus type I) binding protein 1, Tax1 (human T-cell leukemia virus type I) binding protein 2, Tax1-binding protein 2, TAX1BP2FLJ46582, TRXMGC149815, Vac14 homolog (S. cerevisiae) | |
| The epitope recognized by this antibody maps to a region between residue 732 to 782 of human Protein VAC14 homolog using the numbering given in entry Q08AM6.1 (GeneID 55697). | |
| 0.1 mL | |
| Signal Transduction | |
| 55697 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction