missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vaccinia Virus A27L Antibody [mFluor Violet 500 SE] - (Copenhagen), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43776MFV500
This item is not returnable.
View return policy
Description
Vaccinia Virus A27L Polyclonal antibody specifically detects Vaccinia Virus A27L in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Vaccinia Virus A27L | |
| Polyclonal | |
| Western Blot, ELISA | |
| OPG154 | |
| Produced in rabbits immunized with purified, recombinant Vaccinia virus A27L (Copenhagen) (Uniprot#: P20535; Met1-Glu110) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Western Blot | |
| mFluor Violet 500 SE | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction