Learn More
BD Vacutainer™ Glass Cell Preparation Tubes
Product Code. 10381715
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
Closure Color:
Blue/Black
Orange/Green
Additive:
Sodium Citrate/FICOLL
Sodium Heparin
Tube Size:
13 x 100 mm
16 x 125 mm
Unit Size:
Pack of 60
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Additive Tube Size unitSize
10381715 8 mL Blue/Black Sodium Citrate/FICOLL 16 x 125 mm Pack of 60
12917706 8 mL Orange/Green Sodium Heparin 16 x 125 mm Pack of 60
12997696 4 mL Blue/Black Sodium Citrate/FICOLL 13 x 100 mm Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10381715 Supplier BD Supplier No. 362782
Use for the one-step isolation of lymphocytes, monocytes, and plasma. BD™ Vacutainer™ Glass Cell Preparation Tubes allow for collection and processing of whole blood in a closed, sterile tube prepared with sodium citrate or sodium heparin as an anticoagulant.
The BD CPT™ (Cell Preparation Tube) system provides a simple, standardized method for the isolation of Peripheral Blood Mononuclear Cells (PBMCs) from whole blood. In a single process, up to 15 million PBMCs can be isolated within 20 minutes.
  • Improved sample preparation consistency and reproducibility compared to manual gradient separations
  • Faster separations because there is no need to prepare gradients or manipulate the sample
  • Safe isolation of cells with reduced risk of contamination due to the enclosed, sterile tube
Vacutainer™ CPT™ features:
  • Glass molecular diagnostics tubes with see-thru label for easy content viewing
  • Conventional closures are color-coded to easily identify anticoagulant used
  • Density gradient polymer gel with either sodium citrate or sodium heparin anticoagulant
  • Blood draw capacity is 4 or 8mL (4mL tube available in sodium citrate only)
  • Tube is CE marked for in vitro diagnostic use
  • Centrifuging conditions: 1500–1800 g for 15 minutes at 18–25°C
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Product Type Cell Preparation Tube
Additive Sodium Citrate/FICOLL
Material Glass
Sterility Sterile
Closure Type Speckled Conventional
Capacity (Metric) 8 mL
Closure Color Blue/Black
Height (Metric) 125 mm
Tube Size 16 x 125 mm
Includes Polyester gel, density gradient liquid
Label Type Transparent
Diameter (Metric) Outer 16 mm
Certifications/Compliance CE
Show More Show Less
compliance-icons
Product Identifier
  • TUBE CPTCIT GC 16X125 8.0 MLBLCE BL/BLK
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sodium amidotrizoate