Learn More
BD Vacutainer™ Multi-Sample Blood Collection Needles
Product Code. 10715715
Change view
Click to view available options
Gauge:
20 G
22 G
Length (Metric):
38.1 mm
Color:
Black
Yellow
Unit Size:
Pack of 100
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gauge Length (Metric) Color Unit Size
10715715 20 G 38.1 mm Yellow Pack of 100
12957726 22 G 38.1 mm Black Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10715715 Supplier BD Supplier No. 360215
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Can be used with multiple blood collection tubes. BD Vacutainer™ Multi-Sample Blood Collection Needles are sterile, double-ended needles for use with Vacutainer tube holders and vacuum blood collection tubes. Available in 1 in. or 1.5 in. needles of 20, 21 or 22 gauge.
The BD Vacutainer™ system is optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process.
  • Sterile double-ended needle with a sleeve covered safety valve
  • Advanced low-angle bevel design and silicone lubricant coating ensures gentle and smooth vein entry
  • Draws blood directly from the vein into vacuum collection tubes to prevent exposure
  • Can be used with multiple collection tubes after venipuncture
  • Designed for use with BD Vacutainer™ tube holders and vacuum blood collection tubes
  • Luer adapter (MPN 367300) available separately for use with a catheter or hypodermic needle
TRUSTED_SUSTAINABILITY
Specifications
Product Type Multi-sample Blood Collection Needle
Sterility Sterile
Certifications/Compliance CE marked
Color Yellow
Connectors Double-sided
Disposable Single-use
For Use With (Application) Collection of Blood Samples
For Use With (Equipment) Vacutainer Blood Collection Tubes, Tube Holder
Gauge 20 G
Hub Material Polystyrene
Latex Free Yes
Length (English) 1.5 in.
Length (Metric) 38.1 mm
Style Low-angle Bevel
Thickness (Wall) Thin Wall