Learn More
BD Vacutainer™ PET EDTA Tubes
Product Code. 10418334
Change view
Click to view available options
Material:
PET
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
10418334 PET Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10418334 Supplier BD Supplier No. 368856
Edge
Specifications
Product Type Vacutainer EDTA Tube
Additive K2 EDTA
Material PET
Closure Type Hemogard
Capacity (Metric) 3 mL
Closure Color Lavender
Tube Size 13 x 75 mm
For Use With (Application) Whole blood or EDTA plasma sample collection
Label Type Paper