Learn More
Memmert™ Vacuum Oven Thermoshelf
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
Model 200
Model 400
Model 500
Material:
Aluminum
Stainless Steel
Unit Size:
Each
Specifications
Specifications
| Certifications/Compliance | Calibration certificate |
| For Use With (Equipment) | Model 200 |
| Type | Shelf |
| For Use With (Application) | Individual overtemperature protection for each shelf MLOP (Multi-Level-Overtemperature-Control) |
| Includes | Local Temperature Sensing (Pt100, 4-Wire-Circuit) |
| Material | Aluminum |