Learn More
Sartorius™ Vacuum Pump
Brand:  Sartorius™ 16694-2-50-22
1399.99 EUR valid until 2025-12-31
BEST PRICE on Promo! Use promo code "24137" to get your promotional price.
Product Code. 15794088
  • ONLINE EXCLUSIVE PRICE
    € 1855.00
    € 1400.00 / Each
    Save € 455.00
    24% Off
Only null left
Estimated Shipment: 24-04-2026
Specifications
100 mbar
230 V 50 Hz 130 w
130 w
40°C
>22 L/min.
204 mm
Vacuum Pump
230 V
0.9 A
110 mm
7.1 kg
5°C
50 Hz
261 mm
1 bar