Learn More
Welch Ilmvac™ Vacuum Pump Mpc 105 E Diaphragm
Brand:  Welch Ilmvac™ 412421
Additional Details : Weight : 50.00000kg
Product Code. 11891325
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Chemically resistant ultimate pressure <60 mbar 115-230V 50/60Hz 235mm x 140mm x 277mm (w x d x h) pumping speed 0.9/1.0 m3/h Ilmvac