Learn More
Welch Ilmvac™ Vacuum Pump Mpc 110E Diaphragm Chemically Resistant
Shop All Welch Ilmvac Products
Click to view available options
Flow Rate:
20 L/min
Unit Size:
Each
Description
Vacuum Pump, Mpc 110E Diaphragm Chemically Resistant, 90-260V 50/60Hz 235mm x 140mm x 277mm 20L/min flow rate, <50 mbar ultimate vacuum
90-260V 50/60Hz 235mm x 140mm x 277mm 20L/min flow rate, <50 mbar ultimate vacuum
Specifications
Specifications
| Max. Vacuum | 50 Mbar |
| Flow Rate | 20 L/min |
| Pump Type | Vacuum Pump |