Learn More
Welch Ilmvac™ Vacuum pump P 6 E
Brand:  Welch Ilmvac™ 321003
Additional Details : Weight : 19.00000kg
Product Code. 12945134
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
rotary vane 230V 50/60Hz 405mm x 150mm x 215mm (w x d x h) pumping speed 97/110L/min ultimate vacuum 1 x 10-1 mbar Welch-Ilmvac