Learn More
BUCHI Vacuum Pump V-600 for Rotavapor R-220 Pro Industrial Evaporator for multiple benchtop Rotavapors
Product Code. 17224864 Shop All Buchi Products
Click to view available options
Includes:
Secondary Coil Condenser
Secondary Coil Condenser, Woulff Bottle
Secondary Cold Trap Condenser
Secondary Cold Trap Condenser, Woulff Bottle
Woulff Bottle
Unit Size:
Each
Control:
I-300
I-300 Pro
Product Code. 17224864
Brand: BUCHI 11V600120
Only null left
Add to basket
Add to basket
Vacuum Pump V-600 is a chemically resistant 3-stage diaphragm pump with silent and economical operation that makes it suited to work with the R-220 Pro or multiple benchtop Rotavapors.
  • Chemically resistant 3-stage diaphragm pump
  • Quiet and economical operation
  • Optimally suited to work with the R-220 Pro, multiple Rotavapors or SyncorePlus Parallel Evaporation
  • 1.5 mbar final vacuum
  • 3.1 m3/hr. vacuum capacity
Specifications
Max. Vacuum 1.5 mbar
Includes Secondary Cold Trap Condenser, Woulff Bottle
Flow Rate 3.1 m³/hr.
Pump Type Oilless Diaphragm Pump
Voltage 100/240 V