Learn More
Gibertini Elettronica™ VADE 4 Steam Generator
Determine volatile acidity and total sulphurous anhydride (SO2) with this steam generator. Gibertini Elettronica™ VADE 4 Steam Generator is an accessory unit for DEE Distillers.
Brand:  Gibertini Elettronica™ VADE-4
Additional Details : Weight : 9.00000kg
Product Code. 11970625
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Electrically heated boiler 
  • Small size allows for instantaneous and continuous steam generation
  • Reduced pressurized steam volume for increased safety
Specifications
Specifications
Steam Generator
220 V
220 V (-15%/+10%), 15 A
DEE Distillers
130 mm
15 A
405 mm
Determining volatile acidity and total sulphurous anhydride (SO2)
307 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers