missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valacyclovir-d8 HCl (L-valine-d8), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Valaciclovir-D8 (L-valine-D8) Hydrochloride |
| CAS | 1279033-32-7 |
| Molecular Formula | C13 D8 H12 N6 O4 . Cl H |
| Quantity | 0.005 g |
| Synonym | L-Valine-2,3,4,4,4,4',4',4'-d8, 2-[(2-amino-1,6-dihydro-6-oxo-9 H-purin-9-yl)methoxy]ethyl ester, hydrochloride (1:1) (ACI), Valacyclovir-d8 HCl (L-valine-d8), Valaciclovir-D8 (L-valine-D8) hydrochloride |
| InChI Formula | InChI=1 S/C13H20N6O4.ClH/c1-7(2)8(14)12(21)23-4-3-22-6-19-5-16-9-10(19)17-13(15)18-11(9)20;/h5,7-8 H,3-4,6,14H2,1-2H3,(H3,15,17,18,20);1 H/t8-;/m0./s1/i1D3,2D3,7 D,8 D; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])[C@]([2 H])(N)C(=O)OCCOCn1cnc2C(=O)NC(=Nc12)N |
| IUPAC Name | 2-[(2-amino-6-oxo-1 H-purin-9-yl)methoxy]ethyl (2 S)-2-amino-2,3,4,4,4-pentadeuterio-3-(trideuteriomethyl)butanoate;hydrochloride |
| Molecular Weight (g/mol) | 368.846 |
| Formula Weight | 368.181 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction