missing translation for 'onlineSavingsMsg'
Learn More

Valethamate Bromide, Mikromol™

Product Code. 30050977
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30050977

Brand: Mikromol MM3362.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Valethamate Bromide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Valethamate Bromide
CAS 90-22-2
Analyte or Component Names Valethamate Bromide
Shipping Condition Room Temperature
Molecular Formula C19 H32 N O2 . Br
Certifications/Compliance ISO 17025
Synonym Ethanaminium, N,N-diethyl-N-methyl-2-[(3-methyl-1-oxo-2-phenylpentyl)oxy]-, bromide (1:1), Ammonium, diethyl(2-hydroxyethyl)methyl-, bromide, 3-methyl-2-phenylvalerate (8CI), Diethyl(2-hydroxyethyl)methylammonium bromide 3-methyl-2-phenylvalerate (6CI,7CI), Ethanaminium, N,N-diethyl-N-methyl-2-[(3-methyl-1-oxo-2-phenylpentyl)oxy]-, bromide (9CI), Valeric acid, 3-methyl-2-phenyl-, ester with diethyl(2-hydroxyethyl)methylammonium bromide (8CI), 2-Diethylaminoethyl 2-phenyl-3-methylvalerate methyl bromide, 2-Diethylaminoethyl 3-methyl-2-phenylvalerate methylbromide, 3-Methyl-2-phenylvaleric acid 2-diethylaminoethyl ester methyl bromide, 3-Methyl-2-phenylvaleric acid diethyl(2-hydroxyethyl)methylammonium bromide ester, Barespan, Diethyl(2-hydroxyethyl)methylammonium 3-methyl-2-phenylvalerate bromide, Elist, Epidosan, Epidosin, Letamate, Murel, Release V, Resitan, S 78, Valethamate bromide, Velamate, 2-Phenyl-3-methylvaleric acid beta-(diethylamino)ethyl ester bromomethylate
InChI Formula InChI=1S/C19H32NO2.BrH/c1-6-16(4)18(17-12-10-9-11-13-17)19(21)22-15-14-20(5,7-2)8-3;/h9-13,16,18H,6-8,14-15H2,1-5H3;1H/q+1;/p-1
SMILES [Br-].CCC(C)C(C(=O)OCC[N+](C)(CC)CC)c1ccccc1
IUPAC Name diethyl-methyl-[2-(3-methyl-2-phenylpentanoyl)oxyethyl]azanium;bromide
Molecular Weight (g/mol) 386.37
Quantity 100 mg
Formula Weight 385.1616 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.