Learn More
Valproic acid, Lipomed
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Valproic acid
Specifications
Specifications
| Chemical Name or Material | Valproic acid |
| CAS | 99-66-1 |
| Analyte or Component Names | Valproic acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H16 O2 |
| Synonym | Valproic Acid, 2-Propylpentanoic acid, 2-Propylvaleric acid, 4-Heptanecarboxylic acid, 44089, Acetic acid, dipropyl-, DPA, Depakine, Dipropylacetic acid, Ergenyl, Mylproin, NSC 93819, Valproic acid, n-Dipropylacetic acid |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CCCC(CCC)C(=O)O |
| IUPAC Name | 2-propylpentanoic acid |
| Industry Type | Pharmaceutical |
| Show More |