missing translation for 'onlineSavingsMsg'
Learn More

Valrubicin, MedChemExpress

Product Code. 16485848
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16485848

Brand: Medchem Express HY1377210mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Valrubicin is a chemotherapy agent, inhibits TPA- and PDBu-induced PKC activation with IC50s of 0.85 and 1.25 μM, respectively, and has antitumor and antiinflammatory activity.

Valrubicin is a chemotherapy agent, inhibits TPA- and PDBu-induced PKC activation with IC50s of 0.85 and 1.25 μM, respectively, and has antitumor and antiinflammatory activity.

Specifications

Chemical Name or Material Valrubicin
CAS 56124-62-0
Color Red
Purity Grade Notes Research
Molecular Formula C34H36F3NO13
Quantity 10 mg
Synonym AD-32
Solubility Information DMSO : 125 mg/mL (172.74 mM; Need ultrasonic)
SMILES CCCCC(OCC([C@]1(O)CC2=C(C(O)=C3C(C4=C(C(C3=C2O)=O)C=CC=C4OC)=O)[C@@H](O[C@H]5C[C@H](NC(C(F)(F)F)=O)[C@H](O)[C@H](C)O5)C1)=O)=O
Molecular Weight (g/mol) 723.64
Formula Weight 723.64
Percent Purity 98.69%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.