Learn More
Cole-Parmer™ Value Plastics Barbed to Sanitary Adapter
Click to view available options
Material:
Polypropylene
Unit Size:
Each
Description
- These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing.
- Adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow and no particulate build-up.
- No sharp edges in the flow path, cleaning is very easy.
- No BSE documentation required.
- Material meets the requirements for USP Class VI.
- Available with 3/8in. or 1/2in. hose barbs and3/4 in. mini or 1-1/2in. standard sanitary clamp connections.
Specifications
Specifications
| Type | Barbed to Sanitary Adapter, 1/4 in. ID to 1-1/2 in., ADCF-PP |
| Description | Sanitary x Barb |
| Sterilization Method | Ethylene Oxide (ETO) or Gamma |
| Material | Polypropylene |