missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAM1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1366
This item is not returnable.
View return policy
Description
VAM1 Polyclonal specifically detects VAM1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| VAM1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| MAGUK protein p55T, membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6), p55T, protein associated with Lin7 2, VAM-1PALS2VAM1MAGUK p55 subfamily member 6, Veli-associated MAGUK 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Pig and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL, Peptide ELISA Detection limit 1:128000 | |
| NP_057531. | |
| MPP6 | |
| Peptide with sequence QQVLENLTELPSS-C corresponding to N-Terminus according to NP_057531.2. | |
| 0.1 mg | |
| Stem Cell Markers | |
| 51678 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction