missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP-5 Antibody (3E5), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50357AF405
This item is not returnable.
View return policy
Description
VAMP-5 Monoclonal specifically detects VAMP-5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| VAMP-5 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| VAMP5 | |
| GST-mVAMP-5-II and KLH-mVAMP-5-II peptide sequences | |
| 0.1 mL | |
| Membrane Vesicle Markers, Neuronal Cell Markers, Neurotransmission | |
| 10791 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 300000 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| myobrevin, VAMP-5, vesicle-associated membrane protein 5, vesicle-associated membrane protein 5 (myobrevin) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction