missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Swine Influenza A H1N1 Hemagglutinin Antibody (3E9H5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41329H
This item is not returnable.
View return policy
Description
Swine Influenza A H1N1 Hemagglutinin Monoclonal antibody specifically detects Swine Influenza A H1N1 Hemagglutinin in Virus samples. It is validated for ELISA
Specifications
| Swine Influenza A H1N1 Hemagglutinin | |
| Monoclonal | |
| HRP | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
| ELISA | |
| 3E9H5 | |
| PBS | |
| Antibody was raised against a synthetic peptide containing the sequence specific to the novel S-OIV A H1N1 strain hemagglutinin protein. Amino Acid Squence: STSADQQSLYQNAD | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction