missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP-7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 175.00 - € 507.00
Specifications
| Antigen | VAMP-7 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665748
|
Novus Biologicals
NBP2-41183-0.025mg |
0.025 mg |
€ 175.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18730454
|
Novus Biologicals
NBP2-41183 |
0.1 mg |
€ 507.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VAMP-7 Polyclonal specifically detects VAMP-7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| VAMP-7 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ54296, SYBL1FLJ53762, synaptobrevin-like 1, Synaptobrevin-like protein 1, tetanus neurotoxin-insensitive VAMP, Tetanus-insensitive VAMP, Ti-VAMP, TIVAMP, VAMP-7FLJ53045, vesicle-associated membrane protein 7 | |
| VAMP7 | |
| IgG | |
| Affinity Purified | |
| 32 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Endosome Markers | |
| PBS with 0.02% Sodium Azide | |
| 6845 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human VAMP7. The immunogen is located within amino acids 20 - 70 of VAMP7. Amino Acid Squence: AKIPSENNKLTYSHGNYL | |
| Primary | |
| At least three isoforms of VAMP7 are known to exist; this antibody will detect the two longest isoforms. This antibody is predicted to not cross-react with other Ras-related proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title