missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP-7 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32232R
This item is not returnable.
View return policy
Description
VAMP-7 Polyclonal antibody specifically detects VAMP-7 in Human, Mouse samples. It is validated for Western Blot
Specifications
| VAMP-7 | |
| Polyclonal | |
| Western Blot | |
| FLJ54296, SYBL1FLJ53762, synaptobrevin-like 1, Synaptobrevin-like protein 1, tetanus neurotoxin-insensitive VAMP, Tetanus-insensitive VAMP, Ti-VAMP, TIVAMP, VAMP-7FLJ53045, vesicle-associated membrane protein 7 | |
| The immunogen this antibody was made to, maps to a region between residue 105 to 155 of human Vesicle-Associated Membrane Protein 7 using the numbering given in entry NP_005629.1 (GeneID 6845). | |
| 0.1 mL | |
| Endosome Markers | |
| 6845 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction