missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP3/Cellubrevin Antibody (4G9), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97948AF350
This item is not returnable.
View return policy
Description
VAMP3/Cellubrevin Monoclonal specifically detects VAMP3/Cellubrevin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| VAMP3/Cellubrevin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| VAMP3 | |
| Recombinant human cellubrevin (1-77aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA | |
| 4G9 | |
| Western Blot, Flow Cytometry, ELISA | |
| CEBCellubrevin, cellubrevin, SYB3, Synaptobrevin-3, VAMP-3, vesicle-associated membrane protein 3, vesicle-associated membrane protein 3 (cellubrevin) | |
| Mouse | |
| Protein G purified | |
| Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neurotransmission | |
| 9341.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction