missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP3/Cellubrevin Antibody (4G9), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97948APCCY7
This item is not returnable.
View return policy
Description
VAMP3/Cellubrevin Monoclonal specifically detects VAMP3/Cellubrevin in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| VAMP3/Cellubrevin | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| VAMP3 | |
| Recombinant human cellubrevin (1-77aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Anti-human VAMP3 mAb, clone k2A2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human cellubrevin protein. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 4G9 | |
| Flow Cytometry | |
| CEBCellubrevin, cellubrevin, SYB3, Synaptobrevin-3, VAMP-3, vesicle-associated membrane protein 3, vesicle-associated membrane protein 3 (cellubrevin) | |
| Mouse | |
| Protein G purified | |
| Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neurotransmission | |
| 9341.0 | |
| Human, Mouse | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction