Learn More
Invitrogen™ VAMP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592171
Description
Immunogen sequence: MPPKFKRHLN DDDVTGSVKS ERRNLLEDDS DEEEDFFLRG PSGPRFGPRN DKIKHVQNQV DEVIDVMQEN ITKVIERGER LDELQDKSES LSDNATAFSN RSKQLRRQMW WRGCK; Positive Samples: B cells, HeLa, Jurkat, BT-474, NIH/3T3, Mouse brain, Mouse lung, Rat brain; Cellular Location: Golgi apparatus, Single-pass type IV membrane protein, trans-Golgi network membrane.
The vesicle associated membrane proteins (VAMP) or synaptobrevins are calcium binding proteins specific to eukaryotes. VAMPs, along with synaptosomal associated protein of 25 kDa (SNAP-25) and syntaxin, form the core complex of soluble NSF attachment protein receptor (SNARE) proteins that interact with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and alpha-SNAP. These membrane associated proteins play a key role in the regulation of vesicle membrane fusion with the plasma membrane. The Clostridium tetani neurotoxin is a metalloprotease with specificity for VAMP. In Alzheimer's disease, VAMP levels of all isoforms appear to be significantly lowered.
Specifications
| VAMP4 | |
| Polyclonal | |
| Unconjugated | |
| VAMP4 | |
| D1Ertd147e; vamp; VAMP24; VAMP4; VAMP-4; vesicle associated membrane protein 4; vesicle-associated membrane protein 4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 364033, 53330, 8674 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70480, O75379 | |
| VAMP4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-115 of human VAMP4 (NP_0037532). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.