missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAMP8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102921
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total VAMP8.
Involved in the targeting and/or fusion of transport vesicles to their target membrane. Involved for dense-granule secretion in platelets. Plays a role in regulated enzyme secretion in pancreatic acinar cells. Involved in the abscission of the midbody during cell division, which leads to completely separate daughter cells. Involved in the homotypic fusion of early and late endosomes.
Specifications
| VAMP8 | |
| Polyclonal | |
| Unconjugated | |
| VAMP8 | |
| AU041171; Edb; endobrevin; hypothetical protein LOC507309; vamp; VAMP8; VAMP-8; vesicle associated membrane protein 8; vesicle-associated membrane protein 8; vesicle-associated membrane protein 8 (endobrevin); wu:fa18d07; zgc:56588 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8673 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BV40 | |
| VAMP8 | |
| A synthesized peptide derived from human VAMP8(Accession Q9BV40), corresponding to amino acid residues R37-I87. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction