missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vang-like Protein 1/VANGL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
€ 285.00 - € 529.00
Specifications
| Antigen | Vang-like Protein 1/VANGL1 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Knockdown Validated |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18414041
|
Novus Biologicals
NBP1-86990-25ul |
25 μL |
€ 285.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18789573
|
Novus Biologicals
NBP1-86990 |
0.1 mL |
€ 529.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vang-like Protein 1/VANGL1 Polyclonal specifically detects Vang-like Protein 1/VANGL1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
| Vang-like Protein 1/VANGL1 | |
| Polyclonal | |
| Rabbit | |
| Wnt Signaling Pathway | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 81839 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DTESTYSGYSYYSSHSKKSHRQGERTRERHKSPRNKDGRGSEKSVTIQPPTGEPLLGNDSTRTEEVQDDNWGETTTAITGTSEHSISQEDIARISKDMEDSVGLDCKRY | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Knockdown Validated | |
| Unconjugated | |
| RUO | |
| Human | |
| KAI1 C-terminal interacting tetraspanin, Loop-tail protein 2 homolog, LPP2, MGC5338, STB2KITENIN, STBM2, Strabismus 2, Van Gogh-like protein 1, vang (van gogh, Drosophila)-like 1, vang, van gogh-like 1 (Drosophila), vang-like 1 (van gogh, Drosophila), vang-like protein 1 | |
| VANGL1 | |
| IgG | |
| Affinity Purified | |
| Specificity of human Vang-like Protein 1/VANGL1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title