missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vang-like Protein 2/VANGL2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-59736
This item is not returnable.
View return policy
Description
Vang-like Protein 2/VANGL2 Polyclonal specifically detects Vang-like Protein 2/VANGL2 in Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Vang-like Protein 2/VANGL2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| KIAA1215Loop-tail protein 1 homolog, loop-tail-associated protein, LPP1, LTAPVan Gogh-like protein 2, MGC119403, MGC119404, STB1Strabismus 1, STBM, STBM1, vang (van gogh, Drosophila)-like 2, vang, van gogh-like 2 (Drosophila), vang-like 2 (van gogh, Drosophila), vang-like protein 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 57216 | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| Q9ULK5 | |
| VANGL2 | |
| Peptide with sequence C-QKYLRTTKQQPYH corresponding to internal region according to NP_065068.1. | |
| 0.1 mg | |
| Primary | |
| Human, Mouse. Predicted cross-reactivity based on sequence identity: Cow, Zebrafish. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction