missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanilloid R1/TRPV1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71774AF488
This item is not returnable.
View return policy
Description
Vanilloid R1/TRPV1 Polyclonal specifically detects Vanilloid R1/TRPV1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Vanilloid R1/TRPV1 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Q8NER1 | |
| TRPV1 | |
| A genomic peptide made to a C-terminal region of the human TRPV1 protein (within residues 725-839). [Swiss-Prot# Q8NER1] | |
| 0.1 mL | |
| Neuroscience, Phospho Specific, Sensory Systems | |
| 7442 | |
| Store at 4C in the dark. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction