missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VAP-A Polyclonal antibody specifically detects VAP-A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | VAP-A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Formulation | 50mM Sodium Borate |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 125 to 175 of human 33 kDa VAMP-Associated Protein using the numbering given in entry NP_919415.2 (GeneID 9218). |
| Regulatory Status | RUO |
| Gene ID (Entrez) | 9218 |
| Target Species | Human |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?