missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAP-A Antibody (OTI10E10), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74825B
This item is not returnable.
View return policy
Description
VAP-A Monoclonal specifically detects VAP-A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| VAP-A | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| VAPA | |
| Recombinant protein fragment corresponding to amino acids 2-227 of human VAP-A (NP_919415) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI10E10 | |
| Western Blot, Immunohistochemistry | |
| hVAP-33, MGC3745,33 kDa VAMP-associated protein, VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa, VAMP-A, VAP-33, VAP33VAMP (vesicle-associated membrane protein)-associated protein A (33kD), VAP-AVAMP-associated protein A, vesicle-associated membrane protein-associated protein A | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 9218.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction