Learn More
Cole-Parmer™ VapLock™ Activated Media Exhaust Filters
VapLock™ Activated Media Exhaust Filters reduce harmful chemical emissions and odors from solvent containers
Brand:  Cole-Parmer™ 12019-79
Additional Details : Weight : 0.23000kg
 View more versions of this product
Product Code. 17225093
  • € 112.00 / Each
In Stock
to see stock.
Add to basket
Description
Description
Use with indicators to set a replacement schedule
Services
None
Specifications
Specifications
Solvent Safety
Activated Carbon
1/4 in. NPT(F)
Exhaust Filter, 75 g activated carbon, 1/4 in. NPT(F); 1/ea
VapLock Solvent Containment Systems
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers