missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Vapreotide acetate

Product Code. 16474977
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 16474977

Brand: MedChemExpress HYP0061A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vapreotide acetate (RC-160 acetate; BMY-41606 acetate) is a neurokinin-1 (NK1) receptor antagonist, with an IC50 of 330 nM.

Specifications

Purity 98.11%
CAS 849479-74-9
Content And Storage Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 1191.39 g/mol
Product Type Peptides
Quantity 50 mg
Shipping Condition Room temperature
Solubility H2O : 14.29 mg/mL (11.99 mM∣Need ultrasonic)
Synonym RC-160 acetate∣ BMY-41606 acetate
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.